Learn More
Medchemexpress LLC L-allylglycine hydrochloride | 195316-72-4 | MFCD06797042 | 98.0% | 151.59 g/mol | C5H10ClNO2 | 5 G
SDP
Catalog No. 5000300668
Encompass_Preferred
L-Allylglycine hydrochloride is the hydrochloride salt of the amino acid derivative L-allylglycine. It functions as an inhibitor of glutamate decarboxylase (GAD), reducing GABA biosynthesis, and is used in neuroscience and biochemical research. The compound is supplied as a solid powder with reported purity of 98% and is intended for laboratory research use only.
  • Inhibits glutamate decarboxylase (GAD), reducing GABA synthesis.
  • Hydrochloride salt form improves solubility for aqueous studies.
  • High purity (98.0%) suitable for research applications.
  • Available in small-scale sizes for method development and testing.
  • Provided with batch-specific certificate of analysis for traceability.
Catalog No. 50-003-00668 Supplier Medchemexpress LLC Supplier No. HYW0155955G
L-Allylglycine hydrochloride is the hydrochloride salt of the amino acid derivative L-allylglycine. It functions as an inhibitor of glutamate decarboxylase (GAD), reducing GABA biosynthesis, and is used in neuroscience and biochemical research. The compound is supplied as a solid powder with reported purity of 98% and is intended for laboratory research use only.
  • Inhibits glutamate decarboxylase (GAD), reducing GABA synthesis.
  • Hydrochloride salt form improves solubility for aqueous studies.
  • High purity (98.0%) suitable for research applications.
  • Available in small-scale sizes for method development and testing.
  • Provided with batch-specific certificate of analysis for traceability.