Learn More
Cayman Chemical L-AlnIn-d3 100mg
SDP
Catalog No. 502912725
Encompass_Preferred
An internal standard for the quantification of L-alanine by GC- or LC-MS
Catalog No. 50-291-2725 Supplier Cayman Chemical Supplier No. 34833100 MG
An internal standard for the quantification of L-alanine by GC- or LC-MS