Learn More
Cayman Chemical L-Alanine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 63546-27-0, 500 mg
SDP
Catalog No. 502911041
Encompass_Preferred
An internal standard for the quantification of L-alanine by GC- or LC-MS
Catalog No. 50-291-1041 Supplier Cayman Chemical Supplier No. 34833500 MG
An internal standard for the quantification of L-alanine by GC- or LC-MS