Learn More
Cayman Chemical L-Alanine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 63546-27-0, 50 mg
SDP
Catalog No. 502910193
Encompass_Preferred
An internal standard for the quantification of L-alanine by GC- or LC-MS
Catalog No. 50-291-0193 Supplier Cayman Chemical Supplier No. 3483350 MG
An internal standard for the quantification of L-alanine by GC- or LC-MS