Learn More
Cayman Chemical L-Alanine methyl ester (hydrochloride), Purity: ≥95%, CAS: 2491-20-5, 50 g
SDP
Catalog No. 502992004
Encompass_Preferred
An amino acid-containing building block; has been used in the synthesis of AZT nucleotides with anti-HIV activity, as well as anticancer agents
Catalog No. 50-299-2004 Supplier Cayman Chemical Supplier No. 3052750 G
An amino acid-containing building block; has been used in the synthesis of AZT nucleotides with anti-HIV activity, as well as anticancer agents