Learn More
Cayman Chemical L-Alnyl-L-GlutamIn 100g
SDP
Catalog No. 502986616
Encompass_Preferred
A synthetic glutamine dipeptide that can attenuate oxidative stress at doses of at doses of 0.75-1.5 mg/kg; reduces apoptosis in vitro; protects against inflammatory mediators
Catalog No. 50-298-6616 Supplier Cayman Chemical Supplier No. 21809100 G
A synthetic glutamine dipeptide that can attenuate oxidative stress at doses of at doses of 0.75-1.5 mg/kg; reduces apoptosis in vitro; protects against inflammatory mediators