Learn More
Cayman Chemical L-Alanyl-L-Glutamine, Purity: ≥95%, CAS: 39537-23-0, 25 g
SDP
Catalog No. 502986390
Encompass_Preferred
A synthetic glutamine dipeptide that can attenuate oxidative stress at doses of at doses of 0.75-1.5 mg/kg; reduces apoptosis in vitro; protects against inflammatory mediators
Catalog No. 50-298-6390 Supplier Cayman Chemical Supplier No. 2180925 G
A synthetic glutamine dipeptide that can attenuate oxidative stress at doses of at doses of 0.75-1.5 mg/kg; reduces apoptosis in vitro; protects against inflammatory mediators