Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-alpha-amino-beta-ureidopropionic acid | 1483-07-4 | >98.0% | 10 MG
SDP

Catalog No. 5000233664
Encompass_Preferred

L-albizziin is an amino acid analogue used as a glutaminase inhibitor in biochemical and cancer research. It has molecular formula C4H9N3O3 and a molecular weight of 147.13 g/mol.

  • Glutaminase inhibitor for research use.
  • Amino acid analogue suitable for biochemical assays.
  • Cas number 1483-07-4.
  • Molecular formula C4H9N3O3 and molecular weight 147.13 g/mol.
  • Recommended storage: powder at -20°C (long term) or 4°C; in solvent store at -80°C for short term.

Catalog No. 50-002-33664 Supplier Medchemexpress LLC Supplier No. HY12116710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-albizziin is an amino acid analogue used as a glutaminase inhibitor in biochemical and cancer research. It has molecular formula C4H9N3O3 and a molecular weight of 147.13 g/mol.

  • Glutaminase inhibitor for research use.
  • Amino acid analogue suitable for biochemical assays.
  • Cas number 1483-07-4.
  • Molecular formula C4H9N3O3 and molecular weight 147.13 g/mol.
  • Recommended storage: powder at -20°C (long term) or 4°C; in solvent store at -80°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.