Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich L-(-)-Alpha-Amino-Epsilon-Caprolactam Hydrochloride

Catalog No. 216125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-612-5G 5 g
21-612-1G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-612-5G Supplier Sigma Aldrich Supplier No. 216125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

L-(-)-+a-Amino-+a-caprolactam hydrochloride can be used to introduce caprolactam moiety during the total synthesis of bengamides, which are bioactive compounds naturally found in marine sponges.

Specifications

CAS 26081-07-2
Molecular Formula C6H12N2O -+ HCl
Linear Formula C6H12N2O -+ HCl
MDL Number MFCD02683418
Quantity 5 g
Synonym (S)-3-Amino-hexahydro-2-azepinone hydrochloride; L-Lysine lactam hydrochloride
Molecular Weight (g/mol) 164.63 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.