Learn More
Sigma Aldrich Fine Chemicals Biosciences L alpha Hydroxyglutaric ac50MG
Catalog No. 502899288
Encompass
Glutamic acid is metabolized to Hydroxyglutaric acid in an NAD-dependent manner by cell-free extracts of Peptococcus aerogenes. It is formed as an intermediate during glyoxylic acid metabolism in bacteria.
Catalog No. 50-289-9288 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9079050MG
Glutamic acid is metabolized to Hydroxyglutaric acid in an NAD-dependent manner by cell-free extracts of Peptococcus aerogenes. It is formed as an intermediate during glyoxylic acid metabolism in bacteria.