Learn More
Avanti Polar Lipids L-α-phosphatidylethanolamine (Brain, Porcine), 383907-18-4, MFCD02265857, 25 mg
Catalog No. NC1178044
Encompass
Formula Weight: 768.610, Purity >99%, Stability: 3 Months, Storage Temperature: -20°C. Contains ~50% plasmalogen ether lipid.
Catalog No. NC1178044 Supplier Avanti Polar Lipids Supplier No. 840022C25MG
Formula Weight: 768.610, Purity >99%, Stability: 3 Months, Storage Temperature: -20°C. Contains ~50% plasmalogen ether lipid.