Learn More
Abcam L-alpha-Phosphatidylinositol, Phospholipase C substrate, 250MG
Catalog No. 502704877
Encompass_Preferred
MW 867.14 g/mol. Phospholipase C substrate. Phospholipid membrane component. Inositol phosphate precursor. Stimulates vascular Ca²+-dependent protease autolysis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4877 Supplier Abcam Supplier No. AB146179250MG
MW 867.14 g/mol. Phospholipase C substrate. Phospholipid membrane component. Inositol phosphate precursor. Stimulates vascular Ca²+-dependent protease autolysis.
The product is subject to the following: Abcam Restricted Use Statement