Learn More
Medchemexpress LLC L-Amino acid oxidase | 9000-89-9 | 10 MG
SDP
Catalog No. 5000399393
Encompass_Preferred
L-Amino acid oxidase is a homodimeric protein containing flavin adenine dinucleotide. It catalyzes the stereospecific oxidative deamination of L-amino acids to α-keto acids and ammonia.
  • The product is for research use only and not intended for human use.
Catalog No. 50-003-99393 Supplier Medchemexpress LLC Supplier No. HYP298710MG
Add to Cart
Add to Cart
L-Amino acid oxidase is a homodimeric protein containing flavin adenine dinucleotide. It catalyzes the stereospecific oxidative deamination of L-amino acids to α-keto acids and ammonia.
  • The product is for research use only and not intended for human use.