Learn More
Apexbio Technology LLC L-α-Aminoadipic Acid 1118-90-7 1g
SDP
Catalog No. 5000091612
Encompass_Preferred
L-alpha-Aminoadipic Acid (1118-90-7) is a small-molecule inhibitor targeting glutamine synthetase It is designed to competitively inhibit glutamine synthetase activity thereby modulating glutamate metabolism and neurotransmission pathways L-alpha-Aminoadipic Acid exerts its biological activity primarily through stereospecific competitive inhibition of glutamine synthetase In in vitro studies L-alpha-Aminoadipic Acid impacts astrocytic function preceding its gliotoxic effects and modulates kynurenic acid production in hippocampal tissues Based on these pharmacological properties L-alpha-Aminoadipic Acid holds research potential in the investigation of astrocyte function and neurological metabolism within neuroscience and pharmacology fields
Catalog No. 50-000-91612 Supplier Apexbio Technology LLC Supplier No. C54741000
L-alpha-Aminoadipic Acid (1118-90-7) is a small-molecule inhibitor targeting glutamine synthetase It is designed to competitively inhibit glutamine synthetase activity thereby modulating glutamate metabolism and neurotransmission pathways L-alpha-Aminoadipic Acid exerts its biological activity primarily through stereospecific competitive inhibition of glutamine synthetase In in vitro studies L-alpha-Aminoadipic Acid impacts astrocytic function preceding its gliotoxic effects and modulates kynurenic acid production in hippocampal tissues Based on these pharmacological properties L-alpha-Aminoadipic Acid holds research potential in the investigation of astrocyte function and neurological metabolism within neuroscience and pharmacology fields