Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Anserine nitrate | 10030-52-1 | 99.6% | 303.28 | 50 MG
SDP

Catalog No. 5000454789
Encompass_Preferred

L-Anserine nitrate is an endogenous metabolite.

  • Molecular formula: C10H17N5O6
  • Molecular weight: 303.28
  • CAS number: 10030-52-1
  • Purity: 99.64%
  • Appearance: White to off-white solid
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.
  • Solubility in water: 250 mg/mL
  • Solubility in PBS: 50 mg/mL

Catalog No. 50-004-54789 Supplier Medchemexpress LLC Supplier No. HYW01139350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Anserine nitrate is an endogenous metabolite.

  • Molecular formula: C10H17N5O6
  • Molecular weight: 303.28
  • CAS number: 10030-52-1
  • Purity: 99.64%
  • Appearance: White to off-white solid
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.
  • Solubility in water: 250 mg/mL
  • Solubility in PBS: 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.