Learn More
Selleck Chemical LLC L-Anserine nitrate salt S3266-25mg
SDP
Catalog No. 503108782
Encompass_Preferred
L-Anserine nitrate salt a dipeptide found in most animal tissues exhibits antioxidant properties anti-crosslinking and anti-glycation activities L-Anserine protects against neurotoxicity induced by ER-stress inducers
Catalog No. 50-310-8782 Supplier Selleck Chemical LLC Supplier No. S326625MG
L-Anserine nitrate salt a dipeptide found in most animal tissues exhibits antioxidant properties anti-crosslinking and anti-glycation activities L-Anserine protects against neurotoxicity induced by ER-stress inducers