Learn More
TWD Scientific LLC Large Antistatic Polypropylene Weighing Funnel. Case of 160.
SDP
Catalog No. 501173382 Shop All TWD Scientific LLC Products
Encompass_Preferred
Large dispoable antistatic polypropylene weighing funnel, size large. Fits volumetric flask volumes (mL) of 500, 1000. Funnels have a maximum capacity of 20mL solid/10mL liquid. 160/Case
Catalog No. 50-117-3382 Supplier TWD Scientific LLC Supplier No. ASWF3L
Large dispoable antistatic polypropylene weighing funnel, size large. Fits volumetric flask volumes (mL) of 500, 1000. Funnels have a maximum capacity of 20mL solid/10mL liquid. 160/Case