Learn More
TWD Scientific LLC Large Antistatic Polypropylene Weighing Funnel. Pack of 20.
SDP
Catalog No. 501173381 Shop All TWD Scientific LLC Products
Encompass_Preferred
Large dispoable antistatic polypropylene weighing funnel, size large. Fits volumetric flask volumes (mL) of 500, 1000. Funnels have a maximum capacity of 20mL solid/10mL liquid. 20/Pack
Catalog No. 50-117-3381 Supplier TWD Scientific LLC Supplier No. ASWF3LPK
Large dispoable antistatic polypropylene weighing funnel, size large. Fits volumetric flask volumes (mL) of 500, 1000. Funnels have a maximum capacity of 20mL solid/10mL liquid. 20/Pack