Learn More
American Radiolabeled Chemicals Inc L-AP4-3H 250UC
SDP
Catalog No. 5090104514
Encompass
L-AP4-3H 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 183.10, Shipped in Dry Ice, Exclusive, (L-(+)-2-Amino-4-phosphono-2,3-3H-butyric acid)
Catalog No. 50-901-04514 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 0534250UC
Add to Cart
Add to Cart
Request a Quote
L-AP4-3H 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 183.10, Shipped in Dry Ice, Exclusive, (L-(+)-2-Amino-4-phosphono-2,3-3H-butyric acid)