Learn More
Cayman Chemical L-Arabinitol, Purity: ≥95%, CAS: 7643-75-6, 5 g
SDP
Catalog No. 502918546
Encompass_Preferred
An endogenous metabolite; the ratio of D- to L-arabinitol in urine has been used as a marker of Candida infections
Catalog No. 50-291-8546 Supplier Cayman Chemical Supplier No. 388825 G
An endogenous metabolite; the ratio of D- to L-arabinitol in urine has been used as a marker of Candida infections