Learn More
Alkali Scientific L (+) Arabinose, 1 Kg
SDP
Catalog No. 5000660726
Encompass_Preferred
L (+) Arabinose, a monosaccharide sugar, is a carbohydrate reagent available in a 1 kg powder form. This chemical is widely used in laboratory settings for various biochemical applications, including metabolic studies and as a carbon source in microbial culture media. Arabinose is a pentose sugar found in plant cell walls and plays an important role in research involving carbohydrate metabolism and enzymatic reactions. In genetic engineering, it is commonly used in studies involving the arabinose operon, which regulates the metabolism of arabinose in microorganisms. This product is high purity and designed for use in research, testing, and pharmaceutical applications.
Catalog No. 50-006-60726 Supplier Alkali Scientific Supplier No. ARA001CP
L (+) Arabinose, a monosaccharide sugar, is a carbohydrate reagent available in a 1 kg powder form. This chemical is widely used in laboratory settings for various biochemical applications, including metabolic studies and as a carbon source in microbial culture media. Arabinose is a pentose sugar found in plant cell walls and plays an important role in research involving carbohydrate metabolism and enzymatic reactions. In genetic engineering, it is commonly used in studies involving the arabinose operon, which regulates the metabolism of arabinose in microorganisms. This product is high purity and designed for use in research, testing, and pharmaceutical applications.