Learn More
Gold Biotechnology Inc L Arabinose 25 g
SDP
Catalog No. 5000562050
Encompass_Preferred
L-Arabinose is an arabinose isomer widely found in nature while D-arabinose is less common L-Arabinose is a constituent of many biopolymers that make up plant cell walls
Catalog No. 50-005-62050 Supplier Gold Biotechnology Inc Supplier No. A30025
L-Arabinose is an arabinose isomer widely found in nature while D-arabinose is less common L-Arabinose is a constituent of many biopolymers that make up plant cell walls