Learn More
Gold Biotechnology Inc L Arabinose 500 g
SDP
Catalog No. 5000562171
Encompass_Preferred
L-Arabinose is an arabinose isomer widely found in nature while D-arabinose is less common L-Arabinose is a constituent of many biopolymers that make up plant cell walls
Catalog No. 50-005-62171 Supplier Gold Biotechnology Inc Supplier No. A300500
L-Arabinose is an arabinose isomer widely found in nature while D-arabinose is less common L-Arabinose is a constituent of many biopolymers that make up plant cell walls