Learn More
Alkali Scientific L (+) Arabinose, 500 G
SDP
Catalog No. 5000661065
Encompass_Preferred
L (+) Arabinose in a 500 g powder form is a carbohydrate reagent utilized for various biochemical and genetic research applications. Arabinose is commonly used in microbial culture media as a carbon source to study carbohydrate metabolism and enzyme activity. Its role is particularly significant in the arabinose operon, which is widely studied in molecular biology and genetic engineering. Arabinose is also used in the synthesis of arabinogalactans and other polysaccharides. This reagent is often used in experiments involving plant biology, microbiology, and biotechnology, offering high purity and reliability for precise results in laboratory and research settings.
Catalog No. 50-006-61065 Supplier Alkali Scientific Supplier No. ARA002CP
L (+) Arabinose in a 500 g powder form is a carbohydrate reagent utilized for various biochemical and genetic research applications. Arabinose is commonly used in microbial culture media as a carbon source to study carbohydrate metabolism and enzyme activity. Its role is particularly significant in the arabinose operon, which is widely studied in molecular biology and genetic engineering. Arabinose is also used in the synthesis of arabinogalactans and other polysaccharides. This reagent is often used in experiments involving plant biology, microbiology, and biotechnology, offering high purity and reliability for precise results in laboratory and research settings.