Learn More
Gold Biotechnology Inc L-Arabinose 250 g
SDP
Supplier:  Gold Biotechnology Inc A300250
Encompass_Preferred
L-Arabinose is an arabinose isomer widely found in nature, while D-arabinose is less common. L-Arabinose is a constituent of many biopolymers that make up plant cell walls.
Catalog No. 50-153-2732