Learn More
Apexbio Technology LLC L-Arabinose 5328-37-0 500mg
SDP
Catalog No. 5000090890
Encompass_Preferred
L-Arabinose (CAS 5328-37-0) is a pentose monosaccharide derived exclusively from plant sources commonly present as a structural component of plant cell walls Biochemically L-Arabinose functions as a selective noncompetitive inhibitor of intestinal sucrase thereby attenuating sucrose hydrolysis and subsequent postprandial plasma glucose elevations Experimental studies in animal models indicate that dietary administration of L-Arabinose suppresses increases in lipogenic enzyme expression and triacylglycerol levels associated with sucrose intake This compound is thus employed in metabolic and nutritional research to investigate carbohydrate digestion glucose metabolism and lipid regulation
Catalog No. 50-000-90890 Supplier Apexbio Technology LLC Supplier No. B7894500
L-Arabinose (CAS 5328-37-0) is a pentose monosaccharide derived exclusively from plant sources commonly present as a structural component of plant cell walls Biochemically L-Arabinose functions as a selective noncompetitive inhibitor of intestinal sucrase thereby attenuating sucrose hydrolysis and subsequent postprandial plasma glucose elevations Experimental studies in animal models indicate that dietary administration of L-Arabinose suppresses increases in lipogenic enzyme expression and triacylglycerol levels associated with sucrose intake This compound is thus employed in metabolic and nutritional research to investigate carbohydrate digestion glucose metabolism and lipid regulation