Learn More
Medchemexpress LLC L-(+)-arabinose | 5328-37-0 | MFCD00006609 | ≥99.0% | 150.13 g/mol | C5H10O5 | 250 MG
SDP
Catalog No. 5000302572
Encompass_Preferred
L-(+)-Arabinose is a naturally occurring aldopentose supplied as a white to off-white solid for research use. It is commonly used in metabolic studies, carbohydrate research, and microbial induction assays, and is provided with specifications suitable for laboratory applications.
  • High purity (≥99.0%) suitable for biochemical assays.
  • Molecular formula C5H10O5 and molecular weight 150.13 g/mol.
  • White to off-white solid for easy handling and storage.
  • Useful in metabolic and carbohydrate research and as an inducer in bacterial expression systems.
  • Available in multiple laboratory pack sizes for bench-scale experiments.
Catalog No. 50-003-02572 Supplier Medchemexpress LLC Supplier No. 250MGHYW015611R
L-(+)-Arabinose is a naturally occurring aldopentose supplied as a white to off-white solid for research use. It is commonly used in metabolic studies, carbohydrate research, and microbial induction assays, and is provided with specifications suitable for laboratory applications.
  • High purity (≥99.0%) suitable for biochemical assays.
  • Molecular formula C5H10O5 and molecular weight 150.13 g/mol.
  • White to off-white solid for easy handling and storage.
  • Useful in metabolic and carbohydrate research and as an inducer in bacterial expression systems.
  • Available in multiple laboratory pack sizes for bench-scale experiments.