Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-(+)-arabinose | 5328-37-0 | MFCD00135866 | 100.0% | 150.13 g/mol | C5H10O5 | 25 MG
SDP

Catalog No. 5000302856
Encompass_Preferred

L-(+)-Arabinose is an analytical reference standard of the naturally occurring pentose sugar L-arabinose, intended for research and analytical use including assay calibration, method development, and chromatographic reference. It is supplied as a white to off-white crystalline powder with high chemical purity suitable for laboratory applications.

  • Purity 99.95%.
  • Molecular weight 150.13 g/mol.
  • Chemical formula C5H10O5.
  • Supplied as a white to off-white crystalline powder.
  • Pack sizes: 25 mg, 50 mg, 100 mg, 250 mg.
  • Intended for analytical reference, assay calibration, and method validation.
  • Certificate of analysis available for individual batches.

Catalog No. 50-003-02856 Supplier Medchemexpress LLC Supplier No. 25MGHYW015611R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-(+)-Arabinose is an analytical reference standard of the naturally occurring pentose sugar L-arabinose, intended for research and analytical use including assay calibration, method development, and chromatographic reference. It is supplied as a white to off-white crystalline powder with high chemical purity suitable for laboratory applications.

  • Purity 99.95%.
  • Molecular weight 150.13 g/mol.
  • Chemical formula C5H10O5.
  • Supplied as a white to off-white crystalline powder.
  • Pack sizes: 25 mg, 50 mg, 100 mg, 250 mg.
  • Intended for analytical reference, assay calibration, and method validation.
  • Certificate of analysis available for individual batches.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.