Learn More
Abcam L-Arabinose, Ubiquitous naturally occuring saccharide, 25G
Supplier:  Abcam AB14625125G
Encompass_Preferred
MW 150.13 g/mol. Ubiquitous naturally occurring saccharide and constituent of plant polysaccharides. Most bacteria contain an inducible arabinose operon which codes for a series of enzymes and transporters that allows L-arabinose to be used as the sole carbon source in microbial culture.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4798