Learn More
Cayman Chemical L-Arginine-d7 (hydrochloride), Purity: ≥99% deuterated forms (d1-d7), CAS: 204244-77-9, 10 mg
SDP
Catalog No. 502911043
Encompass_Preferred
An internal standard for the quantification of L-arginine by GC- or LC-MS
Catalog No. 50-291-1043 Supplier Cayman Chemical Supplier No. 3483410 MG
An internal standard for the quantification of L-arginine by GC- or LC-MS