Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Argininamide, N-[(phenylmethoxy)carbonyl]glycylglycyl-N-(4-methyl-2-oxo-2H-1-benzopyran-7-yl)- | 66216-78-2 | 99.8% | 579.60 | 10 MG
SDP

Catalog No. 5000426602
Encompass_Preferred

Z-Gly-Gly-Arg-AMC is a thrombin-specific fluorogenic substrate designed for testing thrombin generation in PRP (platelet-rich plasma) and platelet-poor plasma (PPP). It provides specific detection with excitation at 390 nm and emission at 480 nm.

  • Thrombin-specific fluorogenic substrate
  • Supports thrombin generation testing in PRP and PPP
  • Excitation at 390 nm, emission at 480 nm
  • White to off-white solid appearance
  • Purity of 99.80%
  • Soluble in DMSO (100 mg/mL) and H2O (40 mg/mL)
  • Stable as powder for 2 years at -80°C and 1 year at -20°C
  • For research use only

Catalog No. 50-004-26602 Supplier Medchemexpress LLC Supplier No. HYP001910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Gly-Gly-Arg-AMC is a thrombin-specific fluorogenic substrate designed for testing thrombin generation in PRP (platelet-rich plasma) and platelet-poor plasma (PPP). It provides specific detection with excitation at 390 nm and emission at 480 nm.

  • Thrombin-specific fluorogenic substrate
  • Supports thrombin generation testing in PRP and PPP
  • Excitation at 390 nm, emission at 480 nm
  • White to off-white solid appearance
  • Purity of 99.80%
  • Soluble in DMSO (100 mg/mL) and H2O (40 mg/mL)
  • Stable as powder for 2 years at -80°C and 1 year at -20°C
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.