Learn More
Medchemexpress LLC L-Arginine-1-13C hydrochloride | 2483735-41-5 | 211.65 | 1 MG
SDP
Catalog No. 5000389994
Encompass_Preferred
L-Arginine-1-13C (hydrochloride) is the 13C-labeled L-Arginine hydrochloride. L-Arginine hydrochloride is a nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator deficient during sickle cell crisis. This product is for research use only and not sold to patients. It should be stored at -20°C in sealed conditions, away from moisture. When in solvent, store at -80°C for 6 months or -20°C for 1 month, also sealed and away from moisture.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89994 Supplier Medchemexpress LLC Supplier No. HYN0455AS31MG
L-Arginine-1-13C (hydrochloride) is the 13C-labeled L-Arginine hydrochloride. L-Arginine hydrochloride is a nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator deficient during sickle cell crisis. This product is for research use only and not sold to patients. It should be stored at -20°C in sealed conditions, away from moisture. When in solvent, store at -80°C for 6 months or -20°C for 1 month, also sealed and away from moisture.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.