Learn More
Medchemexpress LLC L-Arginine-1-13C hydrochloride | 2483735-41-5 | 211.65 | 50 MG
SDP
Catalog No. 5000390113
Encompass_Preferred
L-Arginine-1-13C (hydrochloride) is the 13C-labeled L-Arginine hydrochloride. L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) is a nitrogen donor for synthesis of nitric oxide, a potent vasodilator that is deficient during times of sickle cell crisis. This product is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-90113 Supplier Medchemexpress LLC Supplier No. HYN0455AS350MG
L-Arginine-1-13C (hydrochloride) is the 13C-labeled L-Arginine hydrochloride. L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) is a nitrogen donor for synthesis of nitric oxide, a potent vasodilator that is deficient during times of sickle cell crisis. This product is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS