Learn More
Medchemexpress LLC L-Arginine-1,2-13C2 (hydrochloride) | 201740-75-2 | 212.65 | 25MG
SDP
Catalog No. 5000389503
Encompass_Preferred
L-Arginine-1,2-13C2 (hydrochloride) is the 13C-labeled L-Arginine hydrochloride, acting as a nitrogen donor for the synthesis of nitric oxide. Nitric oxide is a potent vasodilator that is deficient during times of sickle cell crisis. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • 13C-labeled L-Arginine hydrochloride
  • Nitrogen donor for nitric oxide synthesis
  • Potent vasodilator
  • Used as a tracer for drug development
Catalog No. 50-003-89503 Supplier Medchemexpress LLC Supplier No. 25MGHYN0455AS5
L-Arginine-1,2-13C2 (hydrochloride) is the 13C-labeled L-Arginine hydrochloride, acting as a nitrogen donor for the synthesis of nitric oxide. Nitric oxide is a potent vasodilator that is deficient during times of sickle cell crisis. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • 13C-labeled L-Arginine hydrochloride
  • Nitrogen donor for nitric oxide synthesis
  • Potent vasodilator
  • Used as a tracer for drug development