Learn More
Medchemexpress LLC L-Arginine-1-13C hydrochloride | 2483735-41-5 | 211.65 | 25 MG
SDP
Catalog No. 5000389411
Encompass_Preferred
L-Arginine-1-13C (hydrochloride) is the 13C-labeled L-Arginine hydrochloride. L-Arginine hydrochloride serves as the nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator deficient during sickle cell crises.
  • For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89411 Supplier Medchemexpress LLC Supplier No. 25MGHYN0455AS3
L-Arginine-1-13C (hydrochloride) is the 13C-labeled L-Arginine hydrochloride. L-Arginine hydrochloride serves as the nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator deficient during sickle cell crises.
  • For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs.