Learn More
Abcam L-Arginine, substrate for nitric o x ide synthase, 100MG
Supplier:  Abcam AB120750100MG
Encompass_Preferred
MW 174.2 g/mol, Purity >99%. Precursor to nitric oxide synthesis. Amino acid implicated in neurotransmission, immune defences and cardiovascular endothelial protection. Bioavailability following oral administration is 68%.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1581534