Learn More
Bioworld L-Arginine, 1 kg
Catalog No. 50255283
Encompass_Preferred
A precursor of nitric oxide synthesis that induces vasodilation Synonym: (S)-2-Amino-5-guanidinopentanoic acid Appearance: White crystalline powderBoiling Point : ~367.6 °C at 760 mmHg (Predicted)Density : 1.46 g/cm3HS Code : 29252000IUPAC_Name: (S)-2-Amino-5-guanidinopentanoic acidMelting Point: 222 °CRTECS Number: CF1934200
Catalog No. 50-255-283 Supplier Bioworld Supplier No. 401002683
A precursor of nitric oxide synthesis that induces vasodilation Synonym: (S)-2-Amino-5-guanidinopentanoic acid Appearance: White crystalline powderBoiling Point : ~367.6 °C at 760 mmHg (Predicted)Density : 1.46 g/cm3HS Code : 29252000IUPAC_Name: (S)-2-Amino-5-guanidinopentanoic acidMelting Point: 222 °CRTECS Number: CF1934200