Learn More
Medchemexpress LLC L-Arginine-13C hydrochloride | 94740-43-9 | 211.65 | 25 MG
SDP
Catalog No. 5000389818
Encompass_Preferred
L-Arginine-13C hydrochloride is the 13C-labeled form of L-Arginine hydrochloride. L-Arginine hydrochloride is a nitrogen donor for the synthesis of nitric oxide, a potent vasodilator that is often deficient during sickle cell crisis. This product is intended for research use only and is not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration, for instance, can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89818 Supplier Medchemexpress LLC Supplier No. 25MGHYN0455AS7
L-Arginine-13C hydrochloride is the 13C-labeled form of L-Arginine hydrochloride. L-Arginine hydrochloride is a nitrogen donor for the synthesis of nitric oxide, a potent vasodilator that is often deficient during sickle cell crisis. This product is intended for research use only and is not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration, for instance, can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS