Learn More
Medchemexpress LLC L-Arginine-13C6,15N4 hydrochloride | 202468-25-5 | 220.59 | 1 MG
SDP
Catalog No. 5000389917
Encompass_Preferred
L-Arginine-13C6,15N4 hydrochloride is an isotope-labeled compound, specifically the 13C- and 15N-labeled form of L-Arginine hydrochloride. This compound functions as a nitrogen donor crucial for the synthesis of nitric oxide, a potent vasodilator known to be deficient during sickle cell crises. It is designed for research use, particularly as a tracer and internal standard in quantitative analyses.
  • Can be utilized as a tracer in various analytical methods.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Achieves a high purity of 99.85%.
  • Supplied as a white to off-white solid.
  • Stable for storage at 4°C when sealed and protected from moisture and light.
  • In solvent, stable at -80°C for 6 months or -20°C for 1 month.
  • Highly soluble in H2O at 100 mg/mL (453.33 mM), requiring sonication.
  • Intended for research use only.
Catalog No. 50-003-89917 Supplier Medchemexpress LLC Supplier No. HYN0455AS81MG
L-Arginine-13C6,15N4 hydrochloride is an isotope-labeled compound, specifically the 13C- and 15N-labeled form of L-Arginine hydrochloride. This compound functions as a nitrogen donor crucial for the synthesis of nitric oxide, a potent vasodilator known to be deficient during sickle cell crises. It is designed for research use, particularly as a tracer and internal standard in quantitative analyses.
  • Can be utilized as a tracer in various analytical methods.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Achieves a high purity of 99.85%.
  • Supplied as a white to off-white solid.
  • Stable for storage at 4°C when sealed and protected from moisture and light.
  • In solvent, stable at -80°C for 6 months or -20°C for 1 month.
  • Highly soluble in H2O at 100 mg/mL (453.33 mM), requiring sonication.
  • Intended for research use only.