Learn More
Medchemexpress LLC L-Arginine-13C6,15N4 hydrochloride | 202468-25-5 | 220.59 | 1 MG
SDP
Catalog No. 5000390169
Encompass_Preferred
L-Arginine-13C6,15N4 (hydrochloride) is the 13C- and 15N-labeled L-Arginine hydrochloride. It acts as a nitrogen donor for the synthesis of nitric oxide, a potent vasodilator, which is often deficient during sickle cell crises. This compound is also suitable for use as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Labeled L-Arginine hydrochloride with 13C and 15N isotopes.
  • Acts as a nitrogen donor for nitric oxide synthesis.
  • Potent vasodilator.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis.
Catalog No. 50-003-90169 Supplier Medchemexpress LLC Supplier No. HYN0455AS41MG
L-Arginine-13C6,15N4 (hydrochloride) is the 13C- and 15N-labeled L-Arginine hydrochloride. It acts as a nitrogen donor for the synthesis of nitric oxide, a potent vasodilator, which is often deficient during sickle cell crises. This compound is also suitable for use as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Labeled L-Arginine hydrochloride with 13C and 15N isotopes.
  • Acts as a nitrogen donor for nitric oxide synthesis.
  • Potent vasodilator.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis.