Learn More
Medchemexpress LLC L-Arginine-13C6,15N4 (hydrochloride) | 202468-25-5 | 220.59 | 25 MG
SDP
Catalog No. 5000389440
Encompass_Preferred
L-Arginine-13C6,15N4 (hydrochloride) is the 13C- and 15N-labeled L-Arginine hydrochloride. L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) serves as a nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator and is deficient during sickle cell crises. This product is for research use only and is not sold to patients.
  • 13C- and 15N-labeled L-Arginine hydrochloride
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89440 Supplier Medchemexpress LLC Supplier No. 25MGHYN0455AS8
L-Arginine-13C6,15N4 (hydrochloride) is the 13C- and 15N-labeled L-Arginine hydrochloride. L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) serves as a nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator and is deficient during sickle cell crises. This product is for research use only and is not sold to patients.
  • 13C- and 15N-labeled L-Arginine hydrochloride
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS