Learn More
Medchemexpress LLC L-Arginine-13C6 hydrochloride | 201740-91-2 | 216.62 | 50 MG
SDP
Catalog No. 5000389812
Encompass_Preferred
L-Arginine-13C6 (hydrochloride) is the 13C-labeled L-Arginine hydrochloride. It is a nitrogen donor for the synthesis of nitric oxide, a potent vasodilator deficient during sickle cell crises. This product is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89812 Supplier Medchemexpress LLC Supplier No. HYN0455AS650MG
L-Arginine-13C6 (hydrochloride) is the 13C-labeled L-Arginine hydrochloride. It is a nitrogen donor for the synthesis of nitric oxide, a potent vasodilator deficient during sickle cell crises. This product is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.