Learn More
Medchemexpress LLC L-Arginine-13C6 hydrochloride | 201740-91-2 | MFCD00144558 | 99.0% | 216.62 | 13C6H15ClN4O2 | 10 MG
SDP
Catalog No. 5000295598
Encompass_Preferred
L-Arginine-13C6 hydrochloride is the 13C6-isotopically labeled hydrochloride salt of L-arginine, supplied as a high-purity tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It also serves as a nitrogen donor in research on nitric oxide synthesis.
  • 13C6 isotopic labeling for use as tracer and internal standard.
  • High purity: 99.0%.
  • Hydrochloride salt form for stability and solubility.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Molecular weight 216.62 and CAS 201740-91-2.
Catalog No. 50-002-95598 Supplier Medchemexpress LLC Supplier No. HYN0455AS610MG
L-Arginine-13C6 hydrochloride is the 13C6-isotopically labeled hydrochloride salt of L-arginine, supplied as a high-purity tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It also serves as a nitrogen donor in research on nitric oxide synthesis.
  • 13C6 isotopic labeling for use as tracer and internal standard.
  • High purity: 99.0%.
  • Hydrochloride salt form for stability and solubility.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Molecular weight 216.62 and CAS 201740-91-2.