Learn More
U.S. Pharmacopeia L-Arginine, 74-79-3, MFCD00002635, 200mg
Catalog No. NC9941829
Encompass
Molecular formula C6H14N4O2, Molecular weight 174.2 g/mol, Melting Point 471.2 °F (244 °C), Synonyms: Arginine, L-2-Amino-5-guanidinovaleric acid
Catalog No. NC9941829 Supplier U.S. Pharmacopeia Supplier No. 1042500
Molecular formula C6H14N4O2, Molecular weight 174.2 g/mol, Melting Point 471.2 °F (244 °C), Synonyms: Arginine, L-2-Amino-5-guanidinovaleric acid