Learn More
Medchemexpress LLC L-Arginine-15N2 hydrochloride | 204633-92-1 | 212.65 | 5 MG
SDP
Catalog No. 5000389099
Encompass_Preferred
L-Arginine-15N2 (hydrochloride) is the 15N-labeled L-Arginine (hydrochloride). L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) serves as the nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator that is deficient during sickle cell crises.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89099 Supplier Medchemexpress LLC Supplier No. HYN0455AS5MG
L-Arginine-15N2 (hydrochloride) is the 15N-labeled L-Arginine (hydrochloride). L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) serves as the nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator that is deficient during sickle cell crises.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.