Learn More
Medchemexpress LLC L-arginine-15N2 hydrochloride | 204633-92-1 | MFCD00144557 | 100.0% | 212.65 g·mol⁻¹ | C6H15ClN215N2O2 | 10 MG
SDP
Catalog No. 5000295944
Encompass_Preferred
L-Arginine-15N2 hydrochloride is the 15N-labelled hydrochloride salt of the amino acid L-arginine, supplied as a solid for research use. It is used as a nitrogen donor and tracer in biochemical and metabolic studies, including investigations of nitric oxide synthesis and metabolic flux.
  • 15N-labelled amino acid with reported isotopic purity 99.97%.
  • Solid powder suitable for tracer and analytical applications.
  • Common applications include nitric oxide synthesis studies and metabolic labeling.
  • Store at 4°C, protect from light, and store under inert gas.
  • Provided in small milligram quantities for research use.
Catalog No. 50-002-95944 Supplier Medchemexpress LLC Supplier No. HYN0455AS10MG
L-Arginine-15N2 hydrochloride is the 15N-labelled hydrochloride salt of the amino acid L-arginine, supplied as a solid for research use. It is used as a nitrogen donor and tracer in biochemical and metabolic studies, including investigations of nitric oxide synthesis and metabolic flux.
  • 15N-labelled amino acid with reported isotopic purity 99.97%.
  • Solid powder suitable for tracer and analytical applications.
  • Common applications include nitric oxide synthesis studies and metabolic labeling.
  • Store at 4°C, protect from light, and store under inert gas.
  • Provided in small milligram quantities for research use.