Learn More
Medchemexpress LLC L-arginine-15N2 hydrochloride | 204633-92-1 | MFCD00144557 | 212.65 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0455AS50MG
Encompass_Preferred
L-Arginine-15N2 hydrochloride is the 15N-labeled L-Arginine hydrochloride. L-Arginine hydrochloride is a nitrogen donor for the synthesis of nitric oxide, a potent vasodilator that is deficient during times of sickle cell crisis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88453