Learn More
Medchemexpress LLC L-Arginine-15N4 hydrochloride | 204633-95-4 | MFCD00144559 | 214.64 | 1 MG
SDP
Catalog No. 5000389012
Encompass_Preferred
L-Arginine-15N4 (hydrochloride) is the 15N-labeled L-Arginine hydrochloride. L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) is a nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator deficient during sickle cell crises.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89012 Supplier Medchemexpress LLC Supplier No. HYN0455AS11MG
L-Arginine-15N4 (hydrochloride) is the 15N-labeled L-Arginine hydrochloride. L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) is a nitrogen donor for the synthesis of nitric oxide, which is a potent vasodilator deficient during sickle cell crises.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.