Learn More
Alkali Scientific L Arginine, 25 G
SDP
Catalog No. 5000659760
Encompass_Preferred
L-Arginine, 25 g, is a semi-essential amino acid involved in nitric oxide production, which helps regulate blood flow and vascular function. This amino acid also plays a role in protein synthesis, hormone secretion, and immune system function. The 25g pack is well-suited for smaller-scale research and supplementation applications. L-Arginine is frequently studied for its cardiovascular benefits, such as improving circulation, reducing blood pressure, and enhancing exercise performance. It is also valuable in clinical research related to wound healing, immune support, and metabolic health.
Catalog No. 50-006-59760 Supplier Alkali Scientific Supplier No. LAR006CP
L-Arginine, 25 g, is a semi-essential amino acid involved in nitric oxide production, which helps regulate blood flow and vascular function. This amino acid also plays a role in protein synthesis, hormone secretion, and immune system function. The 25g pack is well-suited for smaller-scale research and supplementation applications. L-Arginine is frequently studied for its cardiovascular benefits, such as improving circulation, reducing blood pressure, and enhancing exercise performance. It is also valuable in clinical research related to wound healing, immune support, and metabolic health.