Learn More
Medchemexpress LLC L-Arginine-7-amido-4-methylcoumarin hydrochloride | 69304-16-1 | 99.23% | 367.83 | 25 MG
SDP
Catalog No. 5000351801
Encompass_Preferred
L-Arginine-7-amido-4-methylcoumarin hydrochloride is a specific substrate for cathepsin H, distinguishing it from cathepsins L and B. This product is intended solely for research purposes.
  • Specific substrate for cathepsin H
  • High purity of 99.23%
  • Solid, white to off-white appearance
  • Soluble in water and DMSO
  • Requires specific storage conditions for stability
Catalog No. 50-003-51801 Supplier Medchemexpress LLC Supplier No. 25MGHY137841
L-Arginine-7-amido-4-methylcoumarin hydrochloride is a specific substrate for cathepsin H, distinguishing it from cathepsins L and B. This product is intended solely for research purposes.
  • Specific substrate for cathepsin H
  • High purity of 99.23%
  • Solid, white to off-white appearance
  • Soluble in water and DMSO
  • Requires specific storage conditions for stability